Skip to Content
Merck
All Photos(1)

Key Documents

792268

Sigma-Aldrich

Eicosanoic-d39 Acid

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)18COOH
CAS Number:
Molecular Weight:
351.77
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

solid

mass shift

M+39

SMILES string

O=C(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H]

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-19H2,1H3,(H,21,22)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2

InChI key

VKOBVWXKNCXXDE-BKDZISOFSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian S W Tordiffe et al.
PloS one, 11(12), e0167608-e0167608 (2016-12-20)
Cheetahs (Acinonyx jubatus) are highly specialised large felids, currently listed as vulnerable on the IUCN red data list. In captivity, they are known to suffer from a range of chronic non-infectious diseases. Although low heterozygosity and the stress of captivity
Eric Tyrode et al.
Nature communications, 11(1), 493-493 (2020-01-26)
Despite the importance of the hydrogen ion in a wide range of biological, chemical, and physical processes, its molecular structure in solution remains lively debated. Progress has been primarily hampered by the extreme diffuse nature of the vibrational signatures of
Arianna Bianchi et al.
Journal of immunology (Baltimore, Md. : 1950), 206(4), 904-916 (2021-01-15)
Age-related chronic inflammation promotes cellular senescence, chronic disease, cancer, and reduced lifespan. In this study, we wanted to explore the effects of a moderate exercise regimen on inflammatory liver disease and tumorigenesis. We used an established model of spontaneous inflammaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service