Skip to Content
Merck
All Photos(1)

Key Documents

777188

Sigma-Aldrich

12-Pentafluorophenoxydodecylphosphonic acid

99% (GC)

Synonym(s):

P-[12-(2,3,4,5,6-Pentafluorophenoxy)dodecyl]phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26F5O4P
CAS Number:
Molecular Weight:
432.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (GC)

form

powder

mp

80-85 °C

SMILES string

OP(O)(=O)CCCCCCCCCCCCOc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C18H26F5O4P/c19-13-14(20)16(22)18(17(23)15(13)21)27-11-9-7-5-3-1-2-4-6-8-10-12-28(24,25)26/h1-12H2,(H2,24,25,26)

InChI key

ULXYEINUTVZCLX-UHFFFAOYSA-N

Application

Organophosphonic acid self-assembled monolayers (SAMs) for nanoelectronics; solid-state lighting; energy generation and nanocomposites

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Randall M Stoltenberg et al.
Langmuir : the ACS journal of surfaces and colloids, 27(1), 445-451 (2010-12-08)
Developing orthogonal surface chemistry techniques that perform at the nanoscale is key to achieving precise control over molecular patterning on surfaces. We report the formation and selective functionalization of alumina nanoparticle arrays generated from block copolymer templates. This new material

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service