Skip to Content
Merck
All Photos(1)

Key Documents

737836

Sigma-Aldrich

Vitamin K3-d8

98 atom % D, 97% (CP)

Synonym(s):

2-(Methyl-d3)-1,4-naphthoquinone-3,5,6,7,8-d5, Menadione-d8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D8O2
CAS Number:
Molecular Weight:
180.23
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

98 atom % D

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

yellow to green

mp

105-107 °C (lit.)

mass shift

M+8

storage temp.

−20°C
−20°C

SMILES string

[2H]c1c([2H])c([2H])c2C(=O)C(=C([2H])C(=O)c2c1[2H])C([2H])([2H])[2H]

InChI

1S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D

InChI key

MJVAVZPDRWSRRC-JGUCLWPXSA-N

Application

Vitamin K3-d8 (or menadione-d8) is a deuterated analog of vitamin K3. It is used as an internal standard for the quantification of vitamin K3 concentration in dietary supplements, fortified foods or within a complex biological matrix.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service