Skip to Content
Merck
All Photos(1)

Key Documents

718033

Sigma-Aldrich

Sucrose solution

NMR reference standard, 2 mM in H2O:D2O (9:1) (99.9 atom % D), sodium azide (trace), sodium chloride 0.25 M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
Molecular Weight:
342.30
UNSPSC Code:
12190000
NACRES:
NA.12

grade

NMR reference standard

form

solution

composition

sodium azide (trace)
sodium chloride, 0.25 M

concentration

2 mM in H2O:D2O (9:1) (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 7 in.
5 mm × 8 in.

Looking for similar products? Visit Product Comparison Guide

Related Categories

Quantity

5 mm O.D. tube contains 0.700 mL.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Matthias Lochner et al.
The Journal of experimental medicine, 205(6), 1381-1393 (2008-05-28)
The nuclear hormone receptor retinoic acid receptor-related orphan receptor gamma t (RORgamma t) is required for the generation of T helper 17 cells expressing the proinflammatory cytokine interleukin (IL)-17. In vivo, however, less than half of RORgamma t(+) T cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service