Skip to Content
Merck
All Photos(1)

Key Documents

70444

Sigma-Aldrich

Tributylmethylammonium chloride

≥98.0% (T)

Synonym(s):

Methyltributylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)3N(Cl)CH3
CAS Number:
Molecular Weight:
235.84
Beilstein:
6300212
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

crystals

SMILES string

[Cl-].CCCC[N+](C)(CCCC)CCCC

InChI

1S/C13H30N.ClH/c1-5-8-11-14(4,12-9-6-2)13-10-7-3;/h5-13H2,1-4H3;1H/q+1;/p-1

InChI key

IPILPUZVTYHGIL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tributylmethylammonium chloride is a quaternary ammonium salt commonly used as a catalyst in the synthesis of ɛ-caprolactone and 1-substituted tetrazoles.

Application

Tributylmethylammonium chloride can be used as a phase transfer catalyst in the synthesis of ɛ-caprolactone by Baeyer-Villiger oxidation of cyclohexanone in the presence of KHSO5 as an oxidizing agent.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New and efficient technique for the synthesis of ?-caprolactone using KHSO5 as an oxidising agent in the presence of a phase transfer catalyst
Baj S, et al
Applied Catalysis A: General, 395(1-2), 49-52 (2011)
One-pot synthesis of 1-substituted 1 H-1, 2, 3, 4-tetrazoles from 2aminothiazoles using tributylmethylammonium chloride as a catalyst
Nagaraju K, et al
Heterocyclic Communications, 23(5), 365-368 (2017)
D M Kou et al.
Se pu = Chinese journal of chromatography, 17(6), 550-552 (2003-01-30)
In this paper, use of tri-n-butylammonium dodecylsulfonate as liquid phase to prepare a capillary column suitable to separate carboxylic acids in a direct injection mode, by the method of dynamic coating with mercury plug, is reported. The results showed that
R H Moseley et al.
The Journal of pharmacology and experimental therapeutics, 276(2), 561-567 (1996-02-01)
Hepatic organic cation transport in vitro, using tetraethylammonium (TEA) as a substrate, consists of at least two steps: sinusoidal uptake is stimulated by an inside-negative membrane potential and canalicular membrane transport is mediated by organic cation:H+ exchange (Moseley et al.
Moon Kyoung Kim et al.
Journal of pharmaceutical sciences, 94(12), 2644-2655 (2005-11-01)
The oral bioavailability of tributylmethyl ammonium (TBuMA), an organic cation (OC), exhibited a dose-dependency (i.e., 17, 27, and 35% at doses of 0.4, 4, or 12 micromol/kg, respectively) in the rat. Relevant mechanisms were investigated in the present study by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service