Skip to Content
Merck
All Photos(3)

Key Documents

700290

Sigma-Aldrich

Cerium(IV) oxide

greener alternative

nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis

Synonym(s):

Ceric oxide, Ceric oxide, ceria

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeO2
CAS Number:
Molecular Weight:
172.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95% trace rare earth metals basis

form

nanopowder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

30 m2/g , BET

particle size

<50 nm (BET)

density

7.13 g/mL at 25 °C (lit.)

bulk density

0.53 g/cm3

greener alternative category

SMILES string

O=[Ce]=O

InChI

1S/Ce.2O

InChI key

CETPSERCERDGAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cerium(IV) oxide (CeO2) is a class of rare earth materials which can be synthesized by a variety of methods which include thermal hydrolysis and hydrothermal method. Its properties include a high refractive index, and dielectric constant.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is intended for Molecular Solar Thermal Energy Storage Systems (MOST) and has been enhanced for energy efficiency. Click here for more information.

Application

CeO2 nanopowder can be used for a variety of applications such as biomedicine, semiconductors, and coating applications.
The additon of CeO2 to CuO/TiO2 increases specific surface area and inhibits sintering and pore size growth at high temperatures.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrothermal synthesis of cerium (IV) oxide
Hirano M and Kato E
Journal of the American Chemical Society, 79(3), 777-780 (1996)
Oxidase-like activity of polymer-coated cerium oxide nanoparticles
Asati A, et al.
Angewandte Chemie (International Edition in English), 48(13), 2308-2312 (2009)
Characterization of cerium (IV) oxide ultrafine particles prepared using reversed micelles
Masui T, et al.
Chemistry of Materials, 9(10), 2197-2204 (1997)
Fungus mediated synthesis of biomedically important cerium oxide nanoparticles
Khan SA and Ahmad A
Materials Research Bulletin, 48(10), 4134-4138 (2013)
Anti-inflammatory Properties of Cerium Oxide Nanoparticles.
Hirst S, et al.
Small, 5(24), 2848-2856 (2009)

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service