Skip to Content
Merck
All Photos(4)

Key Documents

634174

Sigma-Aldrich

Cerium(IV)-zirconium(IV) oxide

greener alternative

nanopowder, <50 nm particle size (BET), 99.0% trace metals basis

Synonym(s):

Cerium zirconate (IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CeO2)·(ZrO2)
CAS Number:
Molecular Weight:
295.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.0% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: cerium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

<50 nm (BET)

density

6.61 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O=[Zr]=O.O=[Ce]=O

InChI

1S/Ce.4O.Zr

InChI key

WKROXIWGJAIPLZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CeO2-ZrO2 composites have a good crystal structure, higher specific surface area, lower band-gap energy, and smaller grain size. It forms p-n heterojunction and is widely used in the field of photocatalysis and oxygen storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for Molecular Solar Thermal Energy Storage Systems (MOST). Click here for more information.

Application

CeO2-ZrO2 can be used as a catalyst for water-splitting reactions and photocatalytic degradation of organic pollutants.

Pt/CeO2–ZrO2 can be used as a catalyst in the dry reformation of methane.

It can also be used to prepare optically passive electrodes for solid-state electrochromic devices.

Features and Benefits

  • Non-toxic
  • High photocatalytic activity
  • Good stability
Optically passive electrode for solid state electrochromic devices.

Legal Information

Product of Engi-Mat Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Hydrogen is one of the most important resources in providing food, fuel, and chemical products for our everyday life. Sustainable catalytic hydrogen production from bioethanol has gained significant attention in recent years due to globally diminishing fossil fuel supplies, which have necessitated the search for new chemical feedstocks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service