Skip to Content
Merck
All Photos(1)

Key Documents

589349

Sigma-Aldrich

Lithocholic acid-2,2,4,4-d4

98 atom % D

Synonym(s):

(3a,5ß)-3-Hydroxycholan-24-oic-2,2,4,4-d4 acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D4H36O3
CAS Number:
Molecular Weight:
380.60
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

mp

184-186 °C (lit.)

mass shift

M+4

SMILES string

[H][C@@]12[C@]([C@](CC([2H])([2H])[C@@H](O)C3([2H])[2H])(C)[C@]3([H])CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(O)=O

InChI

1S/C24H40O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-21,25H,4-14H2,1-3H3,(H,26,27)/t15-,16-,17-,18+,19-,20+,21+,23+,24-/m1/s1/i10D2,14D2

InChI key

SMEROWZSTRWXGI-POXZWENPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aihua Zhao et al.
Scientific reports, 10(1), 11601-11601 (2020-07-16)
Bile acids (BAs) play essential roles in facilitating lipid digestion and absorption in the intestine. Gastric BAs were attributed to abnormal refluxing from duodenal compartments and correlated with the occurrence of gastric inflammation and carcinogenesis. However, the differences in gastric
P Franco et al.
Journal of chromatography. A, 1585, 70-81 (2018-11-30)
Bile acids (BAs) are endogenous steroids involved in the transport of lipids in bile, acting also as molecular signaling hormones. Primary BAs synthesized in the liver undergo several metabolic pathways in the intestine by gut microbiota to produce secondary BAs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service