Skip to Content
Merck
All Photos(1)

Key Documents

556084

Sigma-Aldrich

1-Vinyl cyclohexanol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H10OH
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.478 (lit.)

bp

74 °C/19 mmHg (lit.)

density

0.942 g/mL at 25 °C (lit.)

SMILES string

OC1(CCCCC1)C=C

InChI

1S/C8H14O/c1-2-8(9)6-4-3-5-7-8/h2,9H,1,3-7H2

InChI key

ZXKHOVDDJMJXQP-UHFFFAOYSA-N

General description

1-Vinyl cyclohexanol, also known as 1-vinyl-1-cyclohexanol, is a tertiary allylic alcohol. It can be synthesized from cyclohexanone and vinyl chloride. It can undergo transition-metal-free tandem allylic borylation in the presence of B2pin [bis(pinacolato)diboron], Cs2CO3, THF and MeOH to yield triborated product. Pd-fullerite catalysts have been prepared which effectively catalyzes the hydrogenation of 1-ethynyl-1-cyclohexanol to 1-vinyl-1-cyclohexanol.

Application

1-Vinyl cyclohexanol may be used to synthesize:
  • 1-vinyl-1-cyclohexene
  • 1-vinyl-1-cyclohexylacrylate
  • cyclohexylideneacetaldehyde

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimerization of conjugated cyclodienes.
Suga K, et al.
Canadian Journal of Chemistry, 45(9), 933-937 (1967)
Synthesis of spirocyclic butenolides by ring closing metathesis.
Albrecht U and Langer P.
Tetrahedron, 63(22), 4648-4654 (2007)
One-pot rapid low-cost synthesis of Pd-fullerite catalysts.
Chong LC, et al.
Journal of Materials Chemistry, 18(40), 4808-4813 (2008)
Transition-Metal-Free Borylation of Allylic and Propargylic Alcohols.
Miralles N, et al.
Angewandte Chemie (International Edition in English), 128(13), 4375-4379 (2016)
Lewis Acid Promoted Oxidative Rearrangement of Tertiary Allylic Alcohols with the PhIO/TEMPO System.
Vatele JM.
Synlett, 12, 1785-1788 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service