Skip to Content
Merck
All Photos(1)

Key Documents

544493

Sigma-Aldrich

1-Bromo-2,4-dichlorobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3Cl2
CAS Number:
Molecular Weight:
225.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

26-30 °C (lit.)

SMILES string

Clc1ccc(Br)c(Cl)c1

InChI

1S/C6H3BrCl2/c7-5-2-1-4(8)3-6(5)9/h1-3H

InChI key

ISHYFWKKWKXXPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-2,4-dichlorobenzene is an aryl halide. It can be distinguished from the other bromodichlorobenzene isomers using IR and Raman spectral data. It can undergo Suzuki–Miyaura cross-coupling reaction with arylboronic acids in the presence of different Suzuki catalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeanne L Bolliger et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(13), 4075-4081 (2010-02-27)
Dichloro-bis(aminophosphine) complexes are stable depot forms of palladium nanoparticles and have proved to be excellent Suzuki-Miyaura catalysts. Simple modifications of the ligand (and/or the addition of water to the reaction mixture) have allowed their formation to be controlled. Dichlorobis[1-(dicyclohexylphosphanyl)piperidine]palladium (3)
The 1,3-Diaminobenzene-Derived Aminophosphine Palladium Pincer Complex {C6H3[NHP(piperidinyl)2]2Pd(Cl)}-A Highly Active Suzuki-Miyaura Catalyst with Excellent Functional Group Tolerance.
Bolliger JL and Frech CM.
Advanced Synthesis & Catalysis, 352(6), 1075-1080 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service