Skip to Content
Merck
All Photos(2)

Key Documents

541397

Sigma-Aldrich

10-Bromodecanoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)9CO2H
CAS Number:
Molecular Weight:
251.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reagent type: linker

mp

38-41 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

OC(CCCCCCCCCBr)=O

InChI

1S/C10H19BrO2/c11-9-7-5-3-1-2-4-6-8-10(12)13/h1-9H2,(H,12,13)

InChI key

PGVRSPIEZYGOAD-UHFFFAOYSA-N

General description

10-Bromodecanoic acid can be prepared from 10-bromodecanol via oxidation.

Application

10-Bromodecanoic acid may be employed as an alkylcarboxylate chain source in the preparation of alkylcarboxylate-grafted polyethylenimine. It may also be used in the synthesis of :
  • 10-(methylsulfinyl)decanoic acid
  • 10-cyanodecanoic acid
  • (10-oxo-10-(4-(3-thioxo-3H-1,2-dithiol-5-yl)phenoxy)-decyl)triphenylphosphonium bromide
  • 11-thiastearic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Ayatollahi et al.
The international journal of biochemistry & cell biology, 92, 210-217 (2017-10-17)
RNAi-based gene therapy has been recently considered as a promising approach against cancer. Targeted delivery of drug, gene or therapeutic RNAi-based systems to tumor cells is one of the important issues in order to reduce side effects on normal cells.
Fatemeh Soltani et al.
Artificial cells, nanomedicine, and biotechnology, 45(7), 1356-1362 (2016-11-01)
Poor water solubility of hydrophobic drugs is one of the major problems in pharmaceutical sciences. Herein, in order to address the poor solubility of crocetin, the 30% of primary amines of PAMAM G4 and PPI G4 were alkylated and evaluated
Reza K Oskuee et al.
The journal of gene medicine, 11(10), 921-932 (2009-07-28)
Various strategies have been examined to improve both transfection efficiency and cytotoxicity of polyethylenimine (PEI), a widely used polycationic nonviral gene vector. In the present study, we sought to improve PEI transfection efficiency by combining the osmotic burst mechanism for
M D Rahman et al.
Journal of medicinal chemistry, 31(8), 1656-1659 (1988-08-01)
A variety of analogues of stearic acid in which one of the methylene groups was replaced by a sulfur atom were examined as inhibitors of growth and fatty acid biosynthesis in the trypanosomatid protozoan Crithidia fasciculata. The 8-, 9-, 10-
The synthesis and functional evaluation of a mitochondria-targeted hydrogen sulfide donor,(10-oxo-10-(4-(3-thioxo-3 h-1, 2-dithiol-5-yl) phenoxy) decyl) triphenylphosphonium bromide (Ap39).
Le Trionnaire S, et al.
MedChemComm, 5(6), 728-736 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service