Skip to Content
Merck
All Photos(1)

Key Documents

520462

Sigma-Aldrich

Di(propylene glycol) dimethyl ether, mixture of isomers

≥99.1%

Synonym(s):

Proglyde® DMM, Bis(methoxypropyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC3H6OC3H6OCH3
CAS Number:
Molecular Weight:
162.23
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.1%

refractive index

n20/D 1.408 (lit.)

bp

175 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

SMILES string

COCC(C)OC(C)COC

InChI

1S/C8H18O3/c1-9-5-3-7-11-8-4-6-10-2/h3-8H2,1-2H3

InChI key

KWDCKLXGUZOEGM-UHFFFAOYSA-N

Application

Aprotic solvent for use on proton-sensitive systems such as water-based polyurethane coatings.

Legal Information

Product of Dow Chemical.
Proglyde is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service