Skip to Content
Merck
All Photos(2)

Key Documents

50053

Sigma-Aldrich

Glycidyltrimethylammonium chloride

technical, ≥90% (calc. based on dry substance, AT)

Synonym(s):

(2,3-Epoxypropyl)trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14ClNO
CAS Number:
Molecular Weight:
151.63
Beilstein:
3914931
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (calc. based on dry substance, AT)

impurities

2-4% chlorohydrin
20-25% water

density

1.13 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

[Cl-].C[N+](C)(C)CC1CO1

InChI

1S/C6H14NO.ClH/c1-7(2,3)4-6-5-8-6;/h6H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

PUVAFTRIIUSGLK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Glycidyltrimethylammonium chloride (GTMAC) is a cationizing agent commonly used for starch modification.

Application

Glycidyltrimethylammonium chloride be used as a derivatizing agent to synthesize N-(2-hydroxypropyl)-3-trimethylammonium chitosan chloride (HTCC), a water-soluble chitosan derivative, by reacting with chitosan. HTCC can form nanocomposite films with silver nanoparticles that show good antimicrobial property and optical transparency. Cationic starches can also be prepared by derivatization of starch with GTMAC.

Caution

hydrolysis on storage (approx. 3.5%/month at 20°C)

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakarwan Yostawonkul et al.
Colloids and surfaces. B, Biointerfaces, 149, 301-311 (2016-10-26)
A nanostructure lipid carrier (NLC) composed of solid, and liquid lipid as a core has been developed as a delivery system for hydrophobic drug molecules. The aim of this research was to fabricate an oleoyl-quaternized-chitosan (CS)-coated NLC, where the mucoadhesive
Katariina Solin et al.
Small (Weinheim an der Bergstrasse, Germany), 16(50), e2004702-e2004702 (2020-11-21)
Soft cationic core/shell cellulose nanospheres can deform and interpenetrate allowing their self-assembly into densely packed colloidal nanogel layers. Taking advantage of their water-swelling capacity and molecular accessibility, the nanogels are proposed as a new and promising type of coating material
Xinlu Wang et al.
Frontiers in bioengineering and biotechnology, 7, 360-360 (2020-01-11)
Wound immersion in seawater with high salt, high sodium, and a high abundance of pathogenic bacteria, especially gram-negative bacteria, can cause serious infections and difficulties in wound repair. The present study aimed to prepare a composite hydrogel composed of hyaluronic
Zhao-Xiang Peng et al.
Antimicrobial agents and chemotherapy, 55(2), 860-866 (2010-12-08)
Our previous study (Z. X. Peng et al., Carbohydr. Polym. 81:275-283, 2010) demonstrated that water-soluble quaternary ammonium salts, which are produced by the reaction of chitosan with glycidyl trimethylammonium chloride, provide chitosan derivatives with enhanced antibacterial ability. Because biofilm formation
Le Thi Tuyet Nhung et al.
Polymers, 12(11) (2020-11-21)
An efficient and effective process for the production of high-performance anion exchange membranes (AEMs) is necessary for the commercial application of fuel cells. Therefore, in this study, quaternized poly vinylbenzyl chloride (QVBC) and polysulfone were composited with glycidyltrimethylammonium-chloride-quaternized chitosan (QCS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service