Skip to Content
Merck
All Photos(2)

Key Documents

438243

Sigma-Aldrich

Tetrapropylammonium chloride

98%

Synonym(s):

TPrACl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)4N(Cl)
CAS Number:
Molecular Weight:
221.81
Beilstein:
3567732
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

240-242 °C (lit.)

SMILES string

[Cl-].CCC[N+](CCC)(CCC)CCC

InChI

1S/C12H28N.ClH/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;1H/q+1;/p-1

InChI key

FBEVECUEMUUFKM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetrapropylammonium chloride (TPrACl) can be used as a template in the synthesis of silver nanowires. It can also be used in the synthesis of bis(tetrapropylammonium) hexachlorodicuprate(II) and tetrapropylammonium trichloromercurate(II).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bis (tetrapropylammonium) hexachlorodicuprate (II).
Hu G U O Z H I and Holt E M
Acta Crystallographica Section C, Structural Chemistry, 50(8), 1212-1214 (1994)
Bhavani Prasad Vinjamuri et al.
AAPS PharmSciTech, 18(6), 1925-1935 (2016-11-23)
Spray-dried ipratropium bromide (IPB) microspheres for oral inhalation were engineered using Quality by Design. The interrogation of material properties, process parameters, and critical product quality attributes interplay enabled rational product design. A 2
Far-Infrared and Raman Spectra of Crystalline Tetrapropylammonium Trichloromercurate (II).
Contreras J G and Gloria S
Spectroscopy Letters, 14(7), 553-562 (1981)
Qilong Xie et al.
Analytical and bioanalytical chemistry, 413(2), 577-584 (2020-11-19)
For simultaneous analysis of four fat-soluble tocopherols (α-, β-, γ-, and δ-) in edible oils, an efficient and green method using deep eutectic solvent-based liquid-phase microextraction (DES-LPME) coupled with reversed-phase high-performance liquid chromatography (RP-HPLC) was developed. The DESs formed by
Carmen E Castillo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(15), 4946-4954 (2020-12-23)
Oxoiron(IV) complexes bearing tetradentate ligands have been extensively studied as models for the active oxidants in non-heme iron-dependent enzymes. These species are commonly generated by oxidation of their ferrous precursors. The mechanisms of these reactions have seldom been investigated. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service