Skip to Content
Merck
All Photos(2)

Key Documents

419737

Sigma-Aldrich

6-Methoxyflavone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O3
CAS Number:
Molecular Weight:
252.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

163-165 °C (lit.)

SMILES string

COc1ccc2OC(=CC(=O)c2c1)c3ccccc3

InChI

1S/C16H12O3/c1-18-12-7-8-15-13(9-12)14(17)10-16(19-15)11-5-3-2-4-6-11/h2-10H,1H3

InChI key

XZQLSABETMKIGG-UHFFFAOYSA-N

Gene Information

General description

6-Methoxyflavone is one of the methoxyflavone isolated form Pimelea decora. Synthesis of 6-methoxyflavone from p-dihydroxybenzene has been reported.

Application

6-Methoxyflavone may be employed in the following studies:
  • As internal standard for the analysis of polyphenolic content of in-vitro-cultured chestnut shoots.
  • Synthesis of biflavonoids, rac- and meso-6,6-dimethoxy-2,2-biflavanones.
  • As internal standard for the separation of phenolic compounds in the apricot tissue by HPLC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Shahid et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 95, 1725-1733 (2017-10-01)
Chemotherapy-induced peripheral neuropathy (CIPN) is a major dose limiting side-effect of several commonly used chemotherapeutic agents (such as cisplatin) that profoundly impairs patient quality of life. Unfortunately, neither prophylactic strategies nor symptomatic treatments have proven useful in this condition. Flavonoids
J. L. Fernandez-Lorenzo et al.
Tree physiology, 19(7), 461-466 (2003-03-26)
The phenolic contents of eight in-vitro-cultivated chestnut clones (Castanea sativa Mill. and C. sativa x C. crenata Siebold & Zucc. hybrids) were analyzed both qualitatively and quantitatively. The aim of the work was to identify potential phenolic markers of: (i)
Pilar Errea et al.
Physiologia plantarum, 112(1), 135-141 (2001-04-25)
Graft compatibility has been studied in vitro using callus tissues of apricot (Prunus armeniaca) and different Prunus rootstocks to form scion/rootstock combinations with different degrees of graft compatibility. In these species, incompatibility is manifested by a breakdown of the trees
The constituents of Australian Pimelea species. II. The isolation of unusual flavones from P. simplex and P. decora.
Freeman PW, et al.
Australian Journal of Chemistry, 34(8), 1779-1784 (1981)
Sara Serra et al.
Foods (Basel, Switzerland), 9(10) (2020-10-18)
The rising interest in beneficial health properties of polyphenol compounds in fruit initiated this investigation about biochemical composition in peach mesocarp/exocarp. Biochemical evaluation of phenolic compounds and ascorbic acid were quantified through high-performance liquid chromatography (HPLC) in relation to three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service