Skip to Content
Merck
All Photos(1)

Key Documents

407712

Sigma-Aldrich

Benzylamine

purified by redistillation, ≥99.5%

Synonym(s):

α-Aminotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
741984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.5%

form

liquid

purified by

glass distillation
redistillation

refractive index

n20/D 1.543 (lit.)

bp

184-185 °C (lit.)

mp

10 °C (lit.)

solubility

alcohol: miscible
diethyl ether: soluble
water: miscible

density

0.981 g/mL at 25 °C (lit.)

SMILES string

NCc1ccccc1

InChI

1S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5H,6,8H2

InChI key

WGQKYBSKWIADBV-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)

Looking for similar products? Visit Product Comparison Guide

Application

Benzylamine was used in the following studies:
  • To modify alkanethiol monolayers, via amide and ester formation reactions, for attaching monolayer and submonolayer coverages of specific chemical species onto metal electrodes.
  • As carrier electrolyte for the separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection.
  • To synthesize cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene.
It may be used as a reducing and stabilizing agent to generate reduced graphene oxide (rGO) from graphene oxide.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heuer L.
Ullmann's Encyclopedia of Industrial Chemistry (2006)
Eagleson M.
Concise Encyclopedia Chemistry, 125-125 (1994)
Amide and ester surface attachment reactions for alkanethiol monolayers at gold surfaces as studied by polarization modulation Fourier transform infrared spectroscopy.
Duevel RV and Corn RM.
Analytical Chemistry, 64(4), 227-242 (1992)
A method for the production of reduced graphene oxide using benzylamine as a reducing and stabilizing agent and its subsequent decoration with Ag nanoparticles for enzymeless hydrogen peroxide detection.
Liu S, et al.
Carbon, 49(10), 3158-3164 (2011)
Preparation and characterization of azlactone functionalized polymer supports and their application as scavengers.
Guyomard A, et al.
Eur. Polymer J., 40(10), 2343-2348 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service