Skip to Content
Merck
All Photos(1)

Key Documents

39579

Sigma-Aldrich

2,5-Dimethyl-1,4-benzoquinone

≥98.0%

Synonym(s):

2,5-Dimethyl-2,5-cyclohexadiene-1,4-dione, 2,5-Dimethyl-p-benzoquinone, 2,5-Dimethyl-p-quinone, 2,5-Dimethylbenzoquinone, 2,5-Xyloquinone, 3,6-Dimethyl-p-benzoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O2
CAS Number:
Molecular Weight:
136.15
Beilstein:
2041348
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

mp

123-125 °C

solubility

toluene: soluble 0.5 g/10 mL, clear to faintly turbid, yellow to brown

SMILES string

CC1=CC(=O)C(C)=CC1=O

InChI

1S/C8H8O2/c1-5-3-8(10)6(2)4-7(5)9/h3-4H,1-2H3

InChI key

MYKLQMNSFPAPLZ-UHFFFAOYSA-N

General description

2,5-Dimethyl-1,4-benzoquinone is a quinone derivative. Its reaction with jack bean urease in phosphate buffer, pH 7.8 has been studied. It is present as one of the component of defensive secretion of opilionid Acanthopachylus aculeatus. It has been investigated as inhibitor of jack bean urease in 50mM phosphate buffer, pH 7.0.

Application

2,5-Dimethyl-1,4-benzoquinone may be used in a key step involved in the total synthesis of (−)-cyathin A3.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Floreani et al.
The Journal of pharmacology and experimental therapeutics, 260(2), 468-473 (1992-02-01)
In guinea pig and rat cardiac tissue, redox cycling benzoquinones (2,5-dimethyl-p-benzoquinone and duroquinone) and naphthoquinones (menadione and 2,3-dimethoxy-1,4-naphthoquinone) generated superoxide anion (O2-.) both through one- and two-electron reductions, the generation being significantly greater in guinea pig than in rat tissue.
Wiesława Zaborska et al.
Journal of enzyme inhibition and medicinal chemistry, 17(4), 247-253 (2003-01-18)
1,4-benzoquinone (BQ) and 2,5-dimethyl-1,4-benzoquinone (DMBQ) were studied as inhibitors of jack bean urease in 50 mM phosphate buffer, pH 7.0. The mechanisms of inhibition were evaluated by progress curves studies and steady-state approach to data achieved by preincubation of the
Dale E Ward et al.
Organic letters, 9(15), 2843-2846 (2007-06-22)
The total synthesis of (-)-cyathin A3 is described. The key step involves an unusual enantioselective Diels-Alder reaction of 2,5-dimethyl-1,4-benzoquinone with 2,4-bis(trimethylsilyloxy)-1,3-pentadiene, using Mikami's catalyst [(R)-BINOL + Cl2Ti(OiPr)2 + 4 A mol sieves] modified by addition of Mg and SiO2. Because
J Cao et al.
Biochimica et biophysica acta, 1015(2), 180-188 (1990-02-02)
Upon illumination, a dark-adapted photosynthetic sample shows time-dependent changes in chlorophyll (Chl) a fluorescence yield, known as the Kautsky phenomenon or the OIDPS transient. Based on the differential effects of electron acceptors such as 2,5-dimethyl-p-benzoquinone (DMQ) and 2,6-dichloro-p-benzoquinone (DCBQ) on
S Kumazawa et al.
Journal of bacteriology, 154(1), 185-191 (1983-04-01)
Whole cells of photoanaerobically grown Chromatium sp. strain Miami PBS1071, a marine sulfur purple bacterium, oxidized H2 in the dark through the oxyhydrogen reaction at rates of up to 59 nmol of H2 per mg (dry weight) per min. H2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service