Skip to Content
Merck
All Photos(1)

Key Documents

374539

Sigma-Aldrich

Dimethyl chloromalonate

94%

Synonym(s):

1,3-Dimethyl 2-chloropropanedioate, 2-Chloro-malonic acid dimethyl ester, Chloropropanedioic acid dimethyl ester, Dimethyl α-chloromalonate, Dimethyl 2-chloromalonate, Dimethyl 2-chloropropanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH(COOCH3)2
CAS Number:
Molecular Weight:
166.56
Beilstein:
1768415
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

94%

form

liquid

impurities

4-5% dimethyl dichloromalonate

refractive index

n20/D 1.437 (lit.)

bp

105-106 °C/19 mmHg (lit.)

density

1.305 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(Cl)C(=O)OC

InChI

1S/C5H7ClO4/c1-9-4(7)3(6)5(8)10-2/h3H,1-2H3

InChI key

LNBQBURECUEBKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl chloromalonate (dimethyl 2-chloromalonate) is a dialkyl 2-substituted malonate. Carbanions of dimethyl chloromalonate are reported to react with naphthoquinone derivatives, via vicarious nucleophilic substitution or oxidative nucleophilic substitution of hydrogen processes. Dimethyl chloromalonate is reported to react with hydroquinone and monosubstituted hydroquinones to afford 2-oxido-benzo[b]furan derivatives. Synthesis of dimethyl 2-chloromalonate, via chlorination of dimethyl malonate has been reported.

Application

Dimethyl chloromalonate is suitable for use in the synthesis of 4,5,6-trichloropyrimidine. It may be used in the stereoselective synthesis of highly functionalized nitrocyclopropanes. It may be used in one-pot synthesis of tetra-alkoxycarbonylallylidenetriphenylphosphoranes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vicarious nucleophilic substitution of hydrogen (VNS) in 1, 4-naphthoquinone derivatives-competition between VNS and vinylic nucleophilic substitution (SN V).
Ma?kosza M and Nizamov S.
Tetrahedron, 57(47), 9615-9621 (2001)
An Efficient and Practical Synthesis of Dimethyl 2-chloromalonate.
Babu GR, et al.
Der Pharma Chemica, 3(6) (2011)
The reaction of hydroquinone and monosubstituted hydroquinones with dimethyl chloromalonate.
Renfrew AH, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2382-2386 (1979)
New one-pot synthesis of tetra-alkoxycarbonylallylidenetriphenylphosphoranes.
Yavari I and Baharfar R.
Tetrahedron Letters, 39(9), 1051-1054 (1998)
A preparative method for Synthesis of 4, 5, 6-trichloropyrimidine.
Ostrowski S, et al.
ARKIVOC (Gainesville, FL, United States), 6, 905-908 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service