Skip to Content
Merck
All Photos(2)

Key Documents

347256

Sigma-Aldrich

2,3-Dihydro-2,2-dimethyl-7-benzofuranol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.541 (lit.)

density

1.101 g/mL at 25 °C (lit.)

SMILES string

CC1(C)Cc2cccc(O)c2O1

InChI

1S/C10H12O2/c1-10(2)6-7-4-3-5-8(11)9(7)12-10/h3-5,11H,6H2,1-2H3

InChI key

WJGPNUBJBMCRQH-UHFFFAOYSA-N

General description

2,3-Dihydro-2,2-dimethyl-7-benzofuranol is a 7-phenol metabolite formed during the hydrolysis of carbofuran. 2,3-Dihydro-2,2-dimethyl-7-benzofuranol (ddbfo) reacts with Ti(OiPr)4 or Zr(OEt)4 to yield dimers of [Ti(micro-ddbfo)2(ddbfo)6] and [Zr(ddbfo)3(EtOH)(micro-EtO)]2. Analysis of 2,3-dihydro-2,2-dimethyl-7-benzofuranol in carbofuran by variable-temperature control system using liquid chromatography has been reported.

Application

2,3-Dihydro-2,2-dimethyl-7-benzofuranol was used as starting reagent in the preparation of aminoalkanol derivatives of 2,3-dihydro-2,2-dimethyl-7-benzofuranol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of carbofuran by a pure bacterial culture.
Karns JS, et al.
Pesticide Biochemistry and Physiology, 25(2), 211-217 (1986)
Katarzyna Krauzy-Dziedzic et al.
Dalton transactions (Cambridge, England : 2003), (19)(19), 2620-2626 (2008-04-30)
Reactions of Ti(OiPr)4 or Zr(OEt)4 with 4 equivalents of 2,3-dihydro-2,2-dimethyl-7-benzofuranol (ddbfoH) in toluene gave neutral complexes that in the solid state are dimers of [Ti(micro-ddbfo)2(ddbfo)6] and [Zr(ddbfo)3(EtOH)(micro-EtO)]2 composition. The former could also be conveniently synthesized in a direct reaction of
Inexpensive temperature control system for high-performance liquid chromatography: Application to carbofuran analysis and temperature programming for liquid chromatographic analysis.
Kikta Jr EJ, et al.
Journal of Chromatography A, 138(2), 321-328 (1977)
Synthesis and Structural Characterization of Aminoalkanol Derivatives of 2, 3-Dihydro-2, 2-dimethyl-7-benzofuranol with an Expected beta-Adrenolytic and/or Anxiolytic Activity.
Kossakowski J, et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 57(3), 285-294 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service