Skip to Content
Merck
All Photos(2)

Key Documents

341606

Sigma-Aldrich

Calcium iodate

98%

Synonym(s):

Lautarite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(IO3)2
CAS Number:
Molecular Weight:
389.88
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

solubility

ethanol: insoluble(lit.)
nitric acid: slightly soluble(lit.)

density

4.520 g/mL (lit.)

SMILES string

[Ca++].[O-]I(=O)=O.[O-]I(=O)=O

InChI

1S/Ca.2HIO3/c;2*2-1(3)4/h;2*(H,2,3,4)/q+2;;/p-2

InChI key

UHWJJLGTKIWIJO-UHFFFAOYSA-L

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kaiwen Bai et al.
Poultry science, 96(1), 74-82 (2016-08-04)
This study aimed to investigate the supplemental effects of probiotic Bacillus subtilis fmbJ (BS fmbJ) on growth performance, antioxidant capacity, and meat quality of broiler chickens. A total of 240 day-old male Arbor Acres (AA) broiler chickens were randomly allotted
J A Acosta et al.
Journal of animal science, 95(3), 1243-1252 (2017-04-06)
Rising feed expenditures demand that our industry pursues strategies to lower the cost of production. One option is the adoption of the NE system, although many producers are hesitant to proceed without proof that NE estimates are reliable. The objective
E Merlot et al.
Journal of animal science, 94(2), 563-573 (2016-04-12)
Selection for residual feed intake (RFI), which is used to select animals for feed efficiency, also influences nutrient partitioning between growth and maintenance functions. This study was designed to investigate if selection for reduced RFI can alter the trade-off between
A Zanoletti et al.
Journal of environmental management, 218, 355-362 (2018-04-29)
This work presents a new porous material (SUNSPACE) designed for air particulate matter (PM) capture. It was developed in answer to the European Commission request of an innovative, affordable, and sustainable solution, based on design-driven material, to reduce the concentration
Brooke A Clemmons et al.
PeerJ, 8, e9079-e9079 (2020-05-12)
Overstocking can be a major issue in the dairy cattle industry, leading to negative changes in feeding and resting behavior. Additional stress imposed and alterations in feeding behavior may significantly impact the rumen microbiome. The rumen microbiome is responsible for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service