Skip to Content
Merck
All Photos(1)

Key Documents

333670

Sigma-Aldrich

Tris(trimethylsilyl)phosphine

95%

Synonym(s):

(P(TMS)3), Tris(trimethylsilyl)phosphane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]3P
CAS Number:
Molecular Weight:
250.54
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.502 (lit.)

bp

243-244 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

functional group

phosphine

storage temp.

2-8°C

SMILES string

C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C9H27PSi3/c1-11(2,3)10(12(4,5)6)13(7,8)9/h1-9H3

InChI key

OUMZKMRZMVDEOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tris(trimethylsilyl)phosphine (P(TMS)3) can be used as a precursor for the syntheses of:
  • Indium phosphide (InP) and indium gallium phosphide (InGaP) nanocrystals, type III-V semiconductors.
  • Chiral TMS-phospholane which can be further used to synthesize various bisphospholane ligands for asymmetric catalysis.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Pyr. Liq. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of a new chiral bisphospholane ligand for the Rh (I)-catalyzed enantioselective hydrogenation of isomeric β-acylamido acrylates.
Holz J, et al.
The Journal of Organic Chemistry, 68(5), 1701-1707 (2003)
Yuyuan Wang et al.
ACS applied materials & interfaces, 9(36), 30297-30305 (2017-08-29)
A quantum-dot (QD)-based micelle conjugated with an anti-epidermal growth factor receptor (EGFR) nanobody (Nb) and loaded with an anticancer drug, aminoflavone (AF), has been engineered for EGFR-overexpressing cancer theranostics. The near-infrared (NIR) fluorescence of the indium phosphate core/zinc sulfide shell
Microwave-enhanced reaction rates for nanoparticle synthesis.
Gerbec JA, et al.
Journal of the American Chemical Society, 127(45), 15791-15800 (2005)
Houman Bahmani Jalali et al.
ACS nano, 12(8), 8104-8114 (2018-07-19)
Light-induced stimulation of neurons via photoactive surfaces offers rich opportunities for the development of therapeutic methods and high-resolution retinal prosthetic devices. Quantum dots serve as an attractive building block for such surfaces, as they can be easily functionalized to match
Formation of high quality InP and InAs nanocrystals in a noncoordinating solvent.
Battaglia D and Peng X
Nano Letters, 2(9), 1027-1030 (2002)

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service