Skip to Content
Merck
All Photos(1)

Key Documents

331562

Sigma-Aldrich

Cyclohexanepentanoic acid

98%

Synonym(s):

5-Cyclohexylvaleric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11(CH2)4CO2H
CAS Number:
Molecular Weight:
184.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.4656 (lit.)

bp

126-127 °C/0.5 mmHg (lit.)

mp

16-17 °C (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCCCC1CCCCC1

InChI

1S/C11H20O2/c12-11(13)9-5-4-8-10-6-2-1-3-7-10/h10H,1-9H2,(H,12,13)

InChI key

YMUHUYBRWUUAJF-UHFFFAOYSA-N

Application

Cyclohexanepentanoic acid (5-Cyclohexylvaleric acid) was used as a growth medium for Pseudomonas oleovorans for production of polymers containing cyclohexyl units.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polyesters produced by Pseudomonas oleovorans containing cyclohexyl groups.
Andujar M, et al.
Macromolecules, 30(6), 1611-1615 (1997)
Juan Manuel Gutierrez-Villagomez et al.
Talanta, 162, 440-452 (2016-11-14)
In this study, we report for the first time the efficiency of pentafluorobenzyl bromide (PFBBr) for naphthenic acid (NA) mixtures derivatization, and the comparison in the optimal conditions to the most common NAs derivatization reagents, BF
Dieter Schemeth et al.
Analytica chimica acta, 1038, 182-190 (2018-10-04)
In this study, we focus on isolation and fractionation strategies by solid phase extraction (SPE) for a broad range of environmentally related organic acids. These emerging potential contaminants are primary degradation products of spilled petrogenic compounds but little attention has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service