Skip to Content
Merck
All Photos(2)

Key Documents

326127

Sigma-Aldrich

Indium(III) nitrate hydrate

99.99% trace metals basis

Synonym(s):

Indium trinitrate hydrate, Indium(3+) trinitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In(NO3)3 · xH2O
CAS Number:
Molecular Weight:
300.83 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: indium

impurities

≤150.0 ppm Trace Metal Analysis

SMILES string

[In+3].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/In.3NO3.H2O/c;3*2-1(3)4;/h;;;;1H2/q+3;3*-1;

InChI key

YZZFBYAKINKKFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium(III) nitrate hydrate is an indium based precursor solution, which can be annealed at 500°C and spin coated to form indium oxide thin films.

Application

Indium(III) nitrate hydrate is used in the preparation of indium based substrates for the fabrication of semiconductor devices, such as thin film transistors.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of the synthesis route on the structural properties and shape of the indium oxide (In2O3) nano-particles
Bagheri-Mohagheghi M-M, et al.
Physica E: Low-Dimensional Systems and Nanostructures, 41(10), 1757-1762 (2009)
Influence of precursor concentration on the structural, optical and electrical properties of indium oxide thin film prepared by a sol-gel method
Lau LN, et al.
Applied Surface Science, 345, 355-359 (2015)
High electrical performance of wet-processed indium zinc oxide thin-film transistors
Park K, et al.
IEEE Electron Device Letters, 31(4), 311-313 (2010)
William J Scheideler et al.
ACS applied materials & interfaces, 10(43), 37277-37286 (2018-10-10)
Inorganic transparent metal oxides represent one of the highest performing material systems for thin-film flexible electronics. Integrating these materials with low-temperature processing and printing technologies could fuel the next generation of ubiquitous transparent devices. In this work, we investigate the

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service