Skip to Content
Merck
All Photos(2)

Key Documents

307564

Sigma-Aldrich

Zinc stearate

technical grade

Synonym(s):

Stearic acid zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16COO]2Zn
CAS Number:
Molecular Weight:
632.33
Beilstein:
3919706
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

powder

autoignition temp.

1454 °F

composition

mixture consisting mainly of palmitate (C16) and stearate (C18) salts, 90%
stearate salt : Minimum, ~40% GC

mp

128-130 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Zn]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Zn/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

XOOUIPVCVHRTMJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The thermal decomposition of zinc stearate was studied.

Application

Zinc stearate was used as a reference compound to study the infrared spectra of a colloidal stearate-capped ZnO catalyst substrate for analysing gas adsorption on a colloidal stearate-capped zinc oxide (ZnO) surface. It was also used to prepare the sample suspension injection required for improving the therapeutic treatment for periodontitis.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian P Silverwood et al.
Applied spectroscopy, 68(1), 88-94 (2014-01-11)
Attenuated total reflection Fourier transform infrared (ATR FT-IR) spectroscopy has been applied in situ to study gas adsorption on a colloidal stearate-capped zinc oxide (ZnO) surface. Infrared spectra of a colloidal stearate-capped ZnO catalyst substrate were assigned at room temperature
Enrico Della Gaspera et al.
Journal of the American Chemical Society, 135(9), 3439-3448 (2013-02-12)
We present a new colloidal synthesis of gallium-doped zinc oxide nanocrystals that are transparent in the visible and absorb in the near-infrared. Thermal decomposition of zinc stearate and gallium nitrate after hot injection of the precursors in a mixture of
Yoshihiro Tokudome et al.
Biological & pharmaceutical bulletin, 35(9), 1496-1501 (2012-09-15)
The purpose of this research is improvement of therapeutic treatment for periodontitis by using lipophilic and/or hydrophilic zinc materials. The sample suspension injections were prepared from zinc octanate (C8Zn), zinc stearate (C18Zn), zinc sulfate hepta-hydrate (ZnSO4) and tricalcium phosphate (ZnTCP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service