Skip to Content
Merck
All Photos(3)

Key Documents

306150

Sigma-Aldrich

Dicyclohexyl phthalate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C6H11)2
CAS Number:
Molecular Weight:
330.42
Beilstein:
1889288
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

11.6 (vs air)

vapor pressure

0.1 mmHg ( 150 °C)

Assay

99%

mp

63-67 °C (lit.)

SMILES string

O=C(OC1CCCCC1)c2ccccc2C(=O)OC3CCCCC3

InChI

1S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2

InChI key

VOWAEIGWURALJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dicyclohexyl phthalate (DCHP) is a phthalate ester that is mainly used as a plasticizer in the plastics and packaging industry.

Application

DCHP modified multi-walled carbon nanotubes (MWCNTs) may be used to functionalize screen printed electrodes in biosensor based applications. It may also be used as a reference standard for the determination of dicyclohexyl phthalate in aquatic matrices by ultra-high performance liquid chromatography coupled to hybrid Q-orbitrap mass spectrometry(UHPLC-HRMS).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

356.0 - 374.0 °F

Flash Point(C)

180 - 190 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Double imprinted photoresponsive polymer for simultaneous detection of phthalate esters in plastics.
Gong C, et al.
European Polymer Journal, 108, 295-303 (2018)
Electrochemical acetylcholinesterase biosensor based on multi-walled carbon nanotubes/dicyclohexyl phthalate modified screen-printed electrode for detection of chlorpyrifos.
Chen D, et al.
Journal of Electroanalytical Chemistry, 801, 185-191 (2017)
Targeted quantification and untargeted screening of alkylphenols, bisphenol A and phthalates in aquatic matrices using ultra-high-performance liquid chromatography coupled to hybrid Q-Orbitrap mass spectrometry.
Huysman S, et al.
Analytica Chimica Acta, 801, 185-191 (2018)
S Y Lee et al.
Journal of oral rehabilitation, 25(8), 575-588 (1998-10-22)
The aim of this study was to analyse the leached moieties of dental composites after storage in ethanol and organic acids of plaque and further evaluate the resultant effect on the diametral tensile strength (DTS) of the composites. Three commercial
Eui-Ju Hong et al.
The Journal of reproduction and development, 51(2), 253-263 (2005-05-11)
Phthalates are suspected to disrupt the endocrine system, especially through estrogenic effects. In the present study, we investigated the effects of various phthalates and compared them with those of estrogenic compounds that disrupt the female reproductive system. To assess the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service