Skip to Content
Merck
All Photos(3)

Key Documents

299359

Sigma-Aldrich

Urea-13C

research grade, 99 atom % 13C

Synonym(s):

13C Labeled urea, Carbamide-13C, Carbonyldiamide-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N13CONH2
CAS Number:
Molecular Weight:
61.05
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12

grade

research grade

isotopic purity

99 atom % 13C

Assay

99% (CP)

mp

132-135 °C (lit.)

mass shift

M+1

SMILES string

N[13C](N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)/i1+1

InChI key

XSQUKJJJFZCRTK-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lotterer et al.
Zeitschrift fur Gastroenterologie, 29(11), 590-594 (1991-11-01)
A 13C-urea breath test for detection of Helicobacter pylori infection was validated in 306 patients. Breath samples (four, two or one) were taken at various time points within 30 minutes after a motility inhibiting liquid test meal with citric acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service