Skip to Content
Merck
All Photos(3)

Key Documents

298115

Sigma-Aldrich

Potassium dicyanoaurate(I)

98%

Synonym(s):

Gold (I) potassium cyanide, Potassium gold cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KAu(CN)2
CAS Number:
Molecular Weight:
288.10
Beilstein:
6235525
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

density

3.45 g/mL at 25 °C (lit.)

SMILES string

[K+].N#C[Au-]C#N

InChI

1S/2CN.Au.K/c2*1-2;;/q;;-1;+1

InChI key

OQHPFUBKFKRHKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium dicyanoaurate, also known as potassium gold cyanide. It is a water-soluble salt that is commonly used in gold electroplating, as a precursor for the synthesis of other gold compounds, and in certain chemical processes.

Application

Potassium dicyanoaurate(I) can be used as:     
  • A precursor in the synthesis of gold nanoparticles and nanowires, which are crucial for applications in electronics, photonics, and catalysis     
  • An electrolyte in the potentiostatic deposition of gold nanowires, which are used in electronics and photonics.    
  • A key component in the production of conductive inks and coatings, which are essential for printed circuit boards and other electronic components.
  • A catalyst in certain chemical reactions.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of gold nanowires with controlled crystallographic characteristics.
Karim S, et al.
Applied Physics. A, Materials Science & Processing, 84(4), 403-407 (2006)
Paul Linsdell et al.
The Journal of physiology, 540(Pt 1), 29-38 (2002-04-03)
Lyotropic pseudohalide anions are potentially useful as high affinity probes of Cl(-) channel pores. However, the interaction between these pseudohalides and the cystic fibrosis transmembrane conductance regulator (CFTR) Cl(-) channel have not been described in detail. Here we show that
R Silva et al.
Contact dermatitis, 37(2), 78-81 (1997-08-01)
During 1995, we added gold sodium thiosulfate and potassium dicyanoaurate to the patch test standard series. Our purpose was to compare the reactivity of the gold salts, to determine the frequency of contact allergy to gold in our country, to
G G Graham et al.
Inflammopharmacology, 16(3), 126-132 (2008-06-04)
The aurocyanide anion, Au(CN) (2) (-) , is a human metabolite of several anti-rheumatic gold complexes containing monovalent gold (I) bound to a sulphur ligand. This article reviews some of the chemical and pharmacological properties of this intriguing metabolite, and
Mohammad Fatehi et al.
Biophysical journal, 92(4), 1241-1253 (2006-12-05)
Single channel and macroscopic current recording was used to investigate block of the cystic fibrosis transmembrane conductance regulator (CFTR) Cl(-) channel pore by the permeant anion Au(CN)2(-). Block was 1-2 orders of magnitude stronger when Au(CN)2(-) was added to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service