Skip to Content
Merck
All Photos(1)

Key Documents

290432

Sigma-Aldrich

3,4-Difluorophenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2CO2H
CAS Number:
Molecular Weight:
172.13
Beilstein:
2577768
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

46-50 °C (lit.)

SMILES string

OC(=O)Cc1ccc(F)c(F)c1

InChI

1S/C8H6F2O2/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

YCAKYFIYUHHCKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The separation of five positional isomers from 3,4-difluorophenylacetic acid has been investigated using normal- and reversed-phase HPLC, capillary zone electrophoresis, gas chromatography and supercritical fluid chromatography.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Zhou et al.
Journal of chromatography. A, 866(2), 281-292 (2000-02-12)
The separation of five positional isomers from 3,4-difluorophenylacetic acid was investigated using normal- and reversed-phase high-performance liquid chromatography, capillary zone electrophoresis, gas chromatography and supercritical fluid chromatography. Operating parameters of each technique, such as temperature, type of stationary phase, mobile
Mateusz Daśko et al.
European journal of medicinal chemistry, 128, 79-87 (2017-02-06)
In the present work, we report convenient methods for the synthesis of 3-(4-aminophenyl)-coumarin-7-O-sulfamate derivatives N-acylated with fluorinated analogues of benzoic or phenylacetic acid as steroid sulfatase (STS) inhibitors. The design of these potential STS inhibitors was supported by molecular modeling
Sebastian Demkowicz et al.
Chemical biology & drug design, 87(2), 233-238 (2015-08-19)
In the present work, we report the initial results of our study on a series of 3-phenylcoumarin sulfamate-based compounds containing C-F bonds as novel inhibitors of steroid sulfatase. The new compounds are potent steroid sulfatase inhibitors, possessing more than 10

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service