Skip to Content
Merck
All Photos(1)

Key Documents

270768

Sigma-Aldrich

4-tert-Butylbenzyl bromide

97%

Synonym(s):

α-Bromo-4-(tert-butyl)toluene, 1-Bromomethyl-4-tert-butylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4CH2Br
CAS Number:
Molecular Weight:
227.14
Beilstein:
471674
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.545 (lit.)

bp

93-94 °C/1.5 mmHg (lit.)

mp

8-12 °C (lit.)

density

1.236 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccc(CBr)cc1

InChI

1S/C11H15Br/c1-11(2,3)10-6-4-9(8-12)5-7-10/h4-7H,8H2,1-3H3

InChI key

QZNQSIHCDAGZIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The nucleophilic substitution reaction of 4-tert-butylbenzyl bromide and potassium iodide was carried out in oil-in-water microemulsions based on various surfactants.

Application

4-tert-Butylbenzyl bromide, a hydrophobic reactant, was used to keep the loaded mesoporous material particles under continuous stirring.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomasz Witula et al.
Journal of colloid and interface science, 310(2), 536-545 (2007-03-03)
Mesoporous materials have been investigated as auxiliary agents for organic synthesis comprising reactants with widely different solubility characteristics. The finely divided oxide material was immersed in an aqueous solution of a water-soluble reactant, potassium iodide, and the loaded particles were
Maria Häger et al.
Langmuir : the ACS journal of surfaces and colloids, 20(15), 6107-6115 (2004-07-14)
A nucleophilic substitution reaction between 4-tert-butylbenzyl bromide and potassium iodide has been performed in oil-in-water microemulsions based on various C12Em surfactants, i.e., dodecyl ethoxylate with m number of oxyethylene units. The reaction kinetics was compared with the kinetics of reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service