Skip to Content
Merck
All Photos(1)

Key Documents

257117

Sigma-Aldrich

Phenylmagnesium bromide solution

3.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5MgBr
CAS Number:
Molecular Weight:
181.31
Beilstein:
3588849
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reaction type: Grignard Reaction

concentration

3.0 M in diethyl ether

density

1.134 g/mL at 25 °C

SMILES string

Br[Mg]c1ccccc1

InChI

1S/C6H5.BrH.Mg/c1-2-4-6-5-3-1;;/h1-5H;1H;/q;;+1/p-1

InChI key

ANRQGKOBLBYXFM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:
For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Li et al.
Analytica chimica acta, 884, 106-113 (2015-06-16)
A novel molecularly imprinted electrochemical sensor based on Fe3O4 nanobeads immobilized on graphene (Fe3O4-MIP@RGO) has been developed for detecting 17β-estradiol (17β-E2) in water using reversible addition fragmentation chain transfer (RAFT) polymerization technique. 17β-E2 can be detected by this electrochemical sensor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service