Skip to Content
Merck
All Photos(1)

Key Documents

251631

Sigma-Aldrich

3-Sulfopropyl acrylate potassium salt

Synonym(s):

Potassium 3-(Acryloyloxy)propane-1-sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2)3SO3K
CAS Number:
Molecular Weight:
232.30
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

302 °C (dec.) (lit.)

SMILES string

[K+].[O-]S(=O)(=O)CCCOC(=O)C=C

InChI

1S/C6H10O5S.K/c1-2-6(7)11-4-3-5-12(8,9)10;/h2H,1,3-5H2,(H,8,9,10);/q;+1/p-1

InChI key

VSFOXJWBPGONDR-UHFFFAOYSA-M

Application

The monomer was polymerized to prepare a hydrogel test bed, which was used in a study an in vitro wound infection model. It was polymerized with N-isopropyl acrylamide to prepare thermoresponsive super water absorbent hydrogels, superporous hydrogels (SPH) and SPH based composites.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermoresponsive super water absorbent hydrogels prepared by frontal polymerization of N?isopropyl acrylamide and 3?sulfopropyl acrylate potassium salt
Scognamillo S, et al.
Polymer Science, Series A, 49(5), 1228-1234 (2011)
Thermoresponsive super water absorbent hydrogels prepared by frontal polymerization of N?isopropyl acrylamide and 3?sulfopropyl acrylate potassium salt.
Scognamillo S, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 49(5), 1228-1234 (2011)
Jiehua Lei et al.
ACS applied materials & interfaces, 9(34), 28209-28221 (2017-08-08)
Glycosaminoglycans (GAGs), especially heparin and heparan sulfate (HS), hold great potential for inducing the neural differentiation of embryonic stem cells (ESCs) and have brought new hope for the treatment of neurological diseases. However, the disadvantages of natural heparin/HS, such as
Seung-Min Lim et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(28), 13807-13815 (2019-06-22)
As biological signals are mainly based on ion transport, the differences in signal carriers have become a major issue for the intimate communication between electrical devices and biological areas. In this respect, an ionic device which can directly interpret ionic
In vitro diffusion bed, 3?day repeat challenge `capacity?test for antimicrobial wound dressings
Greenman J, et al.
International Wound Journal, 3(4), 322-329 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service