Skip to Content
Merck
All Photos(1)

Key Documents

250600

Sigma-Aldrich

Tris(dimethylamino)sulfonium difluorotrimethylsilicate

technical grade

Synonym(s):

TAS-F, TASF, Tris(dimethylamino)sulfur trimethylsilyl difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3S[(CH3)3SiF2]
CAS Number:
Molecular Weight:
275.48
Beilstein:
3577020
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

reaction suitability

reagent type: reductant

storage temp.

2-8°C

SMILES string

C[Si-](C)(C)(F)F.CN(C)[S+](N(C)C)N(C)C

InChI

1S/C6H18N3S.C3H9F2Si/c1-7(2)10(8(3)4)9(5)6;1-6(2,3,4)5/h1-6H3;1-3H3/q+1;-1

InChI key

JMGVTLYEFSBAGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(dimethylamino)sulfonium difluorotrimethylsilicate is widely used in organic synthesis as a source of fluoride ion for the synthesis of fluorinated compounds by C-F bond formation. It is also used as a reagent in hydrosilylation, cyanomethylation of ketones and Pd-catalyzed cross-coupling reactions.

Application

Tris(dimethylamino)sulfonium difluorotrimethylsilicate (TASF2SiMe3) can be used as a mild deprotecting reagent for the cleavage of silicon protecting groups.
It can also be used
  • To synthesize deoxyfluoro sugars by reacting with trifluoromethanesulfonyl derivatives of partially protected sugars.
  • As a reagent for the fluorination of hydroxyl-containing compounds.

Other Notes

Remainder tris(dimethylamino)sulfonium bifluoride

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen R Groskreutz et al.
Journal of chromatography. A, 1523, 193-203 (2017-07-20)
On-column solute focusing is a simple and powerful method to decrease the influence of precolumn band spreading and increase the allowable volume injected increasing sensitivity. It relies on creating conditions so that the retention factor, k', is transiently increased during
Utility of tris(dimethylamino)sulphonium difluorotrimethylsilicate (TASF) for the rapid synthesis of deoxyfluoro sugars
Walter A. Szarek, et al.
Journal of the Chemical Society. Chemical Communications, 663-663 (1985)
The rapid synthesis of deoxyfluoro sugars using tris (dimethylamino) sulfonium difluorotrimethylsilicate (TASF)
Doboszewski B, et al.
Canadian Journal of Chemistry, 65(2), 412-419 (1987)
Tris (dimethylamino) sulfonium difluorotrimethylsilicate, a mild reagent for the removal of silicon protecting groups
Scheidt KA, et al.
The Journal of Organic Chemistry, 63(19), 6436-6437 (1998)
Tris (dimethylamino) sulfonium Difluorotrimethylsilicate
RajanBabu TV , et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-12 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service