Skip to Content
Merck
All Photos(1)

Key Documents

249904

Sigma-Aldrich

3,3-Dimethylallyl bromide

technical grade, ~90%

Synonym(s):

1-Bromo-3-methyl-2-butene, Prenyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2Br
CAS Number:
Molecular Weight:
149.03
Beilstein:
1420778
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

~90%

form

liquid

contains

silver wool as stabilizer

refractive index

n20/D 1.489 (lit.)

bp

82-83 °C/150 mmHg (lit.)

density

1.29 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C\C(C)=C\CBr

InChI

1S/C5H9Br/c1-5(2)3-4-6/h3H,4H2,1-2H3

InChI key

LOYZVRIHVZEDMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3-Dimethylallyl bromide has been used in the preparation of:
  • (±)-eldanolide
  • 1-(3,3-dimethylallyl)-L-tryptophan
  • (±)-fumagillin, antibiotic isolated from Aspergillus fumigattus

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A direct synthesis of (?)-eldanolide via the highly regioselective prenylation of 2-trimethylsiloxyfuran.
Jefford CW, et al.
Tetrahedron Letters, 28(9), 949-950 (1987)
A total synthesis of ( )-fumagillin.
E J Corey et al.
Journal of the American Chemical Society, 94(7), 2549-2550 (1972-04-05)
Biosynthesis of aromatic isoprenoids. Part 5. The preparation of 1-(3, 3-dimethylallyl)-L-tryptophan and cyclo-L-aianyl-1-(3, 3-dimethylallyl)-L-tryptophan and their non-incorporation into echinulin.
Grundon MF, et al.
Journal of the Chemical Society. Perkin Transactions 1, 1294-1298 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service