Skip to Content
Merck
All Photos(1)

Key Documents

238627

Sigma-Aldrich

Ethyl isocyanatoacetate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNCH2COOCH2CH3
CAS Number:
Molecular Weight:
129.11
Beilstein:
970503
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

impurities

<5% N-(chloroformyl)glycine ethyl ester

refractive index

n20/D 1.422 (lit.)

bp

67-68 °C/11 mmHg (lit.)

density

1.151 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CN=C=O

InChI

1S/C5H7NO3/c1-2-9-5(8)3-6-4-7/h2-3H2,1H3

InChI key

DUVOZUPPHBRJJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl isocyanatoacetate has been used in the preparation of:
  • 6-(carboxymethylureido)-(±)-nicotine (CMUNic), nicotine immunogen
  • ethyl 8-carbamoyl-4-oxo-3,4-dihydroimidazo[5,1-d]-1,2,3,5-tetrazin-3-ylacetate
  • imidazo[1,2-c]-quinazoline-2,5-(3H,6H)dione

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Facile Synthesis of Imidazo [1, 2-c] quinazoline-2, 5-(3H, 6H) dione.
Papadopoulos EP.
Journal of Heterocyclic Chemistry, 18(3), 515-518 (1981)
Antitumour imidazotetrazines. Part 33. New syntheses of the antitumour drug temozolomide using 'masked'methyl isocyanates.
Malcolm FG.
Journal of the Chemical Society. Perkin Transactions 1, 21, 2783-2787 (1995)
Y Hieda et al.
The Journal of pharmacology and experimental therapeutics, 283(3), 1076-1081 (1998-02-12)
The ability of active immunization to alter nicotine distribution was studied in rats. Animals were immunized with 6-(carboxymethylureido)-(+/-)-nicotine (CMUNic) linked to keyhole limpet hemocyanin (KLH). Antibody titers determined by ELISA, using CMUNic coupled to albumin as the coating antigen, were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service