Skip to Content
Merck
All Photos(1)

Key Documents

237337

Sigma-Aldrich

2-Propylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C6H4NH2
CAS Number:
Molecular Weight:
135.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.547 (lit.)

bp

222-224 °C (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

CCCc1ccccc1N

InChI

1S/C9H13N/c1-2-5-8-6-3-4-7-9(8)10/h3-4,6-7H,2,5,10H2,1H3

InChI key

WKURVXXDGMYSDP-UHFFFAOYSA-N

General description

Electropolymerization of 2-propylaniline in highly acidic medium yields poly(2-propylaniline) film. Polymerization and intercalation of 2-propylaniline into iron (III) oxychloride via in situ polymerization/intercalation process has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (2-propylaniline): an electroactive polymer, soluble in organic medium in the reduced state.
Bidan G, et al.
J. Electroanal. Chem. Interfac. Electrochem., 271(1), 59-68 (1989)
In situ polymerization/intercalation of substituted anilines into iron (III) oxychloride.
Scully SF, et al.
Solid State Ionics, 181(19), 933-938 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service