Skip to Content
Merck
All Photos(2)

Key Documents

230324

Sigma-Aldrich

5-Hexen-1-ol

98%

Synonym(s):

5-Hexenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2(CH2)3CH=CH2
CAS Number:
Molecular Weight:
100.16
Beilstein:
1236458
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

78-80 °C/25 mmHg (lit.)

density

0.834 g/mL at 25 °C (lit.)

SMILES string

OCCCCC=C

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h2,7H,1,3-6H2

InChI key

UIZVMOZAXAMASY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclization to a tetrahydropyran by phenylselenoetherification.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

138.2 °F

Flash Point(C)

59 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Mol. Catal. A: Chem., 272, 288-288 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service