Skip to Content
Merck
All Photos(5)

Key Documents

168149

Sigma-Aldrich

L-Cysteine

97%, for peptide synthesis

Synonym(s):

(R)-2-Amino-3-mercaptopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
121.16
Beilstein:
1721408
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

product name

L-Cysteine, 97%

Assay

97%

form

solid

optical activity

[α]20/D +6.5±1.5°, c = 2 in 5 M HCl

optical purity

ee: 96% (GLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

240 °C (dec.) (lit.)

solubility

hydrochloric acid: 5% in 1N, clear, colorless

application(s)

peptide synthesis

SMILES string

N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1

InChI key

XUJNEKJLAYXESH-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-cysteine is a sulfur-containing non-essential amino acid. Its ability to reduce colitis symptoms is being assessed for potential use in treating inflammatory bowel disease (IBD).
Naturally occurring non-essential amino acid.

Application

L-cysteine can be used as a cysteine source to prepare L-cysteine functionalized gold nanoparticles for use as probes in the colorimetric detection of Hg2+ in aqueous solution.

Biochem/physiol Actions

NMDA glutamatergic receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-cysteine supplementation attenuates local inflammation and restores gut homeostasis in a porcine model of colitis
Kim CJ
Biochim. Biophys. Acta Gen. Subj., 1790(10), 1161-1169 (2009)
Xin Li et al.
Journal of diabetes research, 2020, 9589507-9589507 (2020-02-23)
Chronic wounds are a major global health problem with the presence of biofilm significantly contributing to wound chronicity. Current treatments are ineffective in resolving biofilm and simultaneously killing the bacteria; therefore, effective biofilm-resolving drugs are needed. We have previously shown
L-cysteine functionalized gold nanoparticles for the colorimetric detection of Hg2+ induced by ultraviolet light
Chai F
Nanotechnology, 21(2), 025501-025501 (2009)
Antioxidant effects of sulfur-containing amino acids
Atmaca G
Yonsei Medical Journal, 45, 776-788 (2004)
Tanzila Mukhtar et al.
Scientific reports, 10(1), 4625-4625 (2020-03-15)
Neural stem cells (NSCs) generate neurons of the cerebral cortex with distinct morphologies and functions. How specific neuron production, differentiation and migration are orchestrated is unclear. Hippo signaling regulates gene expression through Tead transcription factors (TFs). We show that Hippo

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service