Skip to Content
Merck
All Photos(1)

Key Documents

158844

Sigma-Aldrich

2-Methylthio-2-imidazoline hydriodide

99%

Synonym(s):

2-Methylsulfanyl-4,5-dihydro-1H-imidazole hydrogen iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2S · HI
CAS Number:
Molecular Weight:
244.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

144-146 °C (lit.)

SMILES string

I[H].CSC1=NCCN1

InChI

1S/C4H8N2S.HI/c1-7-4-5-2-3-6-4;/h2-3H2,1H3,(H,5,6);1H

InChI key

PZZRSEUDGCFXIH-UHFFFAOYSA-N

Application

2-Methylthio-2-imidazoline hydriodide was used in the preparation of starting materials required for the synthesis of 2-amino1-(2-Imidazolin-2-yl)-2-imidazoline.
Reactant for synthesis of:
Aurora and epidermal growth factor receptor kinase inhibitors
Spiro-piperidine inhibitors against influenza A virus
Galegine analogues for aided weight loss in mice
Guanidinyl pyrrolidines as bifunctional catalysts for enantioselective conjugate additions to cyclic enones
Carbonic anhydrase inhibitors
Small molecule CXCR4 antagonists

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-(2-Imidazolin-2-yl)-2-imidazolines. I. Structure of Jaffe's base and the chemistry of related compounds.
Wittekind RR, et al.
The Journal of Organic Chemistry, 38(9), 1641-1645 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service