Skip to Content
Merck
All Photos(2)

Key Documents

158194

Sigma-Aldrich

2,2′-Dithiobis(5-nitropyridine)

96%

Synonym(s):

Bis(5-nitro-2-pyridyl) disulfide, DTNP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N4O4S2
CAS Number:
Molecular Weight:
310.31
Beilstein:
305413
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

155-157 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(SSc2ccc(cn2)[N+]([O-])=O)nc1

InChI

1S/C10H6N4O4S2/c15-13(16)7-1-3-9(11-5-7)19-20-10-4-2-8(6-12-10)14(17)18/h1-6H

InChI key

ROUFCTKIILEETD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2′-Dithiobis(5-nitropyridine) is an aromatic disulphide.

Application

2,2′-Dithiobis(5-nitropyridine) was employed:
  • as cysteine-activating reagent to study the NMR of G protein-coupled receptors
  • in the deprotection assays for protected selenocysteine-containing peptides
  • for deprotecting p-methoxybenzyl groups and acetamidomethyl groups from the side-chains of cysteine and selenocysteine
  • to remove the p-methoxybenzyl protecting group from cysteine and selenocysteine side-chains

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Qi et al.
Nature communications, 11(1), 1262-1262 (2020-03-11)
Binding of biomolecules to crystal surfaces is critical for effective biological applications of crystalline nanomaterials. Here, we present the modulation of exposed crystal facets as a feasible approach to enhance specific nanocrystal-biomolecule associations for improving cellular targeting and nanomaterial uptake.
Katharine M Harris et al.
Journal of peptide science : an official publication of the European Peptide Society, 13(2), 81-93 (2006-10-13)
We present here a simple method for deprotecting p-methoxybenzyl groups and acetamidomethyl groups from the side-chains of cysteine and selenocysteine. This method uses the highly elecrophilic, aromatic disulfides 2,2'-dithiobis(5-nitropyridine) (DTNP) and 2,2'-dithiodipyridine (DTP) dissolved in TFA to effect removal of
Uncoupling and energy transfer inhibition of photophosphorylation by sulfhydryl reagents.
J V Moroney et al.
The Journal of biological chemistry, 255(14), 6670-6674 (1980-07-25)
Effects of oxidants on membrane potential, K+ and Ca2+ currents of mouse pancreatic B-cells.
P Krippeit-Drews et al.
Advances in experimental medicine and biology, 426, 355-359 (1997-01-01)
K Yamaoka et al.
Pflugers Archiv : European journal of physiology, 440(2), 207-215 (2000-07-18)
The effects of sulfhydryl (SH) reagents on the L-type Ca current (ICa) were studied in frog ventricular myocytes using the whole-cell patch-clamp method. Methanethiosulfonate ethylammonium (MTSEA+) was found to enter the cell through the membrane and cause a remarkable increase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service