Skip to Content
Merck
All Photos(1)

Key Documents

124931

Sigma-Aldrich

(R)-(−)-Carvone

98%

Synonym(s):

(−)-Carvone, (R)-5-Isopropenyl-2-methyl-2-cyclohexenone, p-Mentha-6,8-dien-2-one, Carvol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2206714
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.2 (vs air)

vapor pressure

0.4 mmHg ( 20 °C)

Assay

98%

form

liquid

optical activity

[α]20/D −61°, neat

refractive index

n20/D 1.497 (lit.)

bp

227-230 °C (lit.)

density

0.959 g/mL at 25 °C (lit.)

SMILES string

CC(=C)[C@@H]1CC=C(C)C(=O)C1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m1/s1

InChI key

ULDHMXUKGWMISQ-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(−)-Carvone is a monoterpenoid. Its ozonolysis in the gaseous phase has been proposed. Aldehydes (formaldehyde) and biradicals were obtained as the major products. It participates in the diastereoselective synthesis of homochiral octalones.

Application

(R)-(−)-Carvone may be employed as starting reagent for the synthesis of enantiopure (R)-(+)-3-methyl-6-isopropenyl-cyclohept-3-enone-1, a useful intermediate formed during the synthesis of terpenoids. It may be employed as starting reagent for the synthesis of differently protected (4S,6R,7R)-trihydroxy-1-octyne derivatives.
Chiral starting material.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas-phase ozonolysis of the monoterpenoids (S)-(+)-carvone,(R)-(-)-carvone,(-)-carveol, geraniol and citral.
Nunes FMN, et al.
Atmospheric Environment, 39(40), 7715-7730 (2005)
Liebigs Ann. Chem., 403-403 (1993)
Bryostatin: A novel asymmetric synthesis of the C27-C34 fragment starting from (R)-carvone as chiral template.
De Brabander J, et al.
Tetrahedron Asymmetry, 8(11), 1721-1724 (1997)
Yoshiyuki Yamada et al.
Neuron, 93(5), 1198-1212 (2017-02-28)
Sensory information is translated into ensemble representations by various populations of projection neurons in brain circuits. The dynamics of ensemble representations formed by distinct channels of output neurons in diverse behavioral contexts remains largely unknown. We studied the two output
An improved synthesis of homochiral octalones from (-)-carvone.
Tenius BSM, et al.
Tetrahedron Asymmetry, 4(4), 633-636 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service