Skip to Content
Merck
All Photos(2)

Key Documents

12255

Sigma-Aldrich

1,2-Benzisoxazole

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO
CAS Number:
Molecular Weight:
119.12
Beilstein:
2154
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

refractive index

n20/D 1.561 (lit.)
n20/D 1.563

bp

90-92 °C/15 mmHg (lit.)

density

1.174 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

c1ccc2oncc2c1

InChI

1S/C7H5NO/c1-2-4-7-6(3-1)5-8-9-7/h1-5H

InChI key

KTZQTRPPVKQPFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Benzisoxazole derivative zonisamide is a novel antiepileptic drug and is effective for the treatment of partial seizures. 1,2-Benzisoxazole is a potential substrates of rabbit liver aldehyde oxidase.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D H Peters et al.
Drugs, 45(5), 760-787 (1993-05-01)
Zonisamide is a 1,2 benzisoxazole derivative and the first agent of this chemical class to be developed as an antiepileptic drug. It has shown activity in various animal models of epilepsy, and although a detailed mode of action awaits clarification
Min Bao et al.
International journal of biological macromolecules, 137, 537-544 (2019-06-25)
Studies on endo-inulinases from yeast are scarce, compared to those from other microbial sources. In this study, a novel endo-inulinase from Lipomyces starkeyi NRRL Y-11557 was identified, expressed in its soluble form, and characterized its physicochemically properties, together with its
Nan-Sook Hong et al.
Nature communications, 9(1), 3900-3900 (2018-09-27)
Developments in computational chemistry, bioinformatics, and laboratory evolution have facilitated the de novo design and catalytic optimization of enzymes. Besides creating useful catalysts, the generation and iterative improvement of designed enzymes can provide valuable insight into the interplay between the
W Gristwood et al.
Xenobiotica; the fate of foreign compounds in biological systems, 18(8), 949-954 (1988-08-01)
1. Twelve oxygen and sulphur azaheterocycles were studied as potential substrates of rabbit liver aldehyde oxidase. Only benzoxazole and 1,2-benzisoxazole were found to be substrates. 2. Nine of the compounds inhibited the oxidation of quinazoline by aldehyde oxidase and in
Avneet Kaur et al.
Archiv der Pharmazie, 351(6), e1800008-e1800008 (2018-05-10)
A series of N-(2-(3,5-dimethoxyphenyl)benzoxazole-5-yl)benzamide derivatives (3am) was synthesized and evaluated for their in vitro inhibitory activity against COX-1 and COX-2. The compounds with considerable in vitro activity (IC50  < 1 μM) were evaluated in vivo for their anti-inflammatory potential by the carrageenan-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service