Skip to Content
Merck
All Photos(2)

Key Documents

112194

Sigma-Aldrich

Hexachloro-1,3-butadiene

96%

Synonym(s):

Perchlorobutadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl2=CClCCl=CCl2
CAS Number:
Molecular Weight:
260.76
Beilstein:
1766570
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.2 mmHg ( 20 °C)

Assay

96%

form

liquid

refractive index

n20/D 1.555 (lit.)

bp

210-220 °C (lit.)

mp

−22-−19 °C (lit.)

density

1.665 g/mL at 25 °C (lit.)

SMILES string

Cl\C(Cl)=C(Cl)/C(Cl)=C(\Cl)Cl

InChI

1S/C4Cl6/c5-1(3(7)8)2(6)4(9)10

InChI key

RWNKSTSCBHKHTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexachloro-1,3-butadiene was used to study the effect of anethol dithiolthione (ADT) on hexachloro-1,3-butadiene (HCBD)-induced nephrotoxicity in rats.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Bouthillier et al.
Toxicology and applied pharmacology, 139(1), 177-185 (1996-07-01)
Administration of anethol dithiolthione (ADT) to rodents can afford protection against some chemically induced toxicities. The aim of the present study was to assess the effects of ADT on hexachloro-1,3-butadiene (HCBD)-induced nephrotoxicity in the rat and to determine the mechanism
M T Li et al.
Bioresource technology, 99(15), 6878-6884 (2008-03-14)
A bacterium (strain HL1) capable of growing with hexachlorobutadiene (HCBD) as sole carbon and energy sources was isolated from a mixture of soil contaminated with HCBD and activated sludge obtained from petrochemical plant wastewater treatment plant by using enrichment culture.
Geoffrey Barnes et al.
Social science & medicine (1982), 55(12), 2227-2241 (2002-11-01)
This paper contributes to the literature on community response to the announcement of well-established chemical contamination close to their homes. It describes a study of residents' views of chemical contamination on a close and long-standing community in the context of
Chon-Lin Lee et al.
Chemosphere, 58(11), 1503-1516 (2005-02-08)
Forty sediments were analyzed for chlorobenzenes (CBs) and hexachlorobutadiene (HCBD) to investigate their pollution topography along the Kaohsiung coast, Taiwan. Maximum CB concentrations found, varied from 15.4ng/gdw for 1,2,3-TCB to 56.8ng/gdw for 1,2,4-TCB, while higher HCBD concentrations were detected among
Aubrey Swain et al.
Journal of applied toxicology : JAT, 31(4), 366-377 (2011-01-25)
Hexachloro-1:3-butadiene (HCBD) causes kidney injury specific to the pars recta of the proximal tubule. In the present studies, injury to the nephron was characterized at 24 h following a single dose of HCBD, using a range of quantitative urinary measurements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service