Skip to Content
Merck
All Photos(1)

Key Documents

108049

Sigma-Aldrich

Ethyl phenylacetate

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2COOC2H5
CAS Number:
Molecular Weight:
164.20
Beilstein:
509140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

refractive index

n20/D 1.497 (lit.)

bp

229 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)Cc1ccccc1

InChI

1S/C10H12O2/c1-2-12-10(11)8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

DULCUDSUACXJJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl phenylacetate is a less toxic, greener solvent. It is non-mutagenic.

Application

Ethyl phenylacetate was used to study the solvent-based self-healing of epoxy materials.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acid catalysis in amines. II. The catalytic effect of various butylammonium salts on the aminolysis of ethyl phenylacetate in anhydrous n-butylamine.
Glasoe PK, et al.
Journal of the American Chemical Society, 61(9), 2387-2389 (1939)
Lara Tat et al.
Journal of agricultural and food chemistry, 55(13), 5205-5212 (2007-05-29)
Interest in high-quality and peculiar products is a recent trend in the enological field; for this reason, production of wines from autochthonous vine varieties is requested by consumers. Aglianico wine from the Italian region "Basilicata" is an example of a
Full Recovery of Fracture Toughness Using a Nontoxic Solvent-Based Self-Healing System.
Caruso MM, et al.
Advances in Functional Materials, 18(13), 1898-1904 (2008)
Lulu Wang et al.
Journal of agricultural and food chemistry, 68(6), 1666-1677 (2020-01-21)
A potent unpleasant aroma presenting a pickle-like off-odor in Moutai-aroma type Baijiu was studied by comparative aroma extract dilution analysis (AEDA). Two Moutai-aroma type Baijiu samples, one having no off-odor (A) and the other one presenting pickle-like off-odor (B), were
Eduardo Coelho et al.
Food chemistry, 323, 126826-126826 (2020-04-27)
During beer ageing, endogenous barrel microbes grow spontaneously and transform wort/beer composition, being Dekkera bruxellensis and Saccharomyces cerevisiae among the main contributors to the chemical and sensory profile of aged beer. This work aims at the application of multi-starter cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service