Skip to Content
Merck
All Photos(3)

Key Documents

107395

Sigma-Aldrich

Phenylacetaldehyde

≥90%

Synonym(s):

α-Tolyaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CHO
CAS Number:
Molecular Weight:
120.15
Beilstein:
385791
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023706
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90%

refractive index

n20/D 1.535 (lit.)

bp

195 °C

mp

−10 °C (lit.)

solubility

H2O: slightly soluble
alcohol: soluble
diethyl ether: soluble

density

1.027 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

O=CCc1ccccc1

InChI

1S/C8H8O/c9-7-6-8-4-2-1-3-5-8/h1-5,7H,6H2

InChI key

DTUQWGWMVIHBKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylacetaldehyde was used in a study to analyse the role of plant derived volatile chemicals on foraging of 1st instar Helicoverpa armigera.

Biochem/physiol Actions

Phenylacetaldehyde is an insect attractant and can be used in blacklight trap for pests. It is constituent of floral scent. It is an intermediate in a variety of biochemical pathways.

Other Notes

Contains varying amounts of poly(styrene oxide)

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

154.4 °F - (External MSDS)

Flash Point(C)

68 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of two plant-derived volatiles in the foraging movement of 1st instar Helicoverpa armigera (Hubner): time to stop and smell the flowers.
Perkins LE, et al.
Arthropod-Plant Interactions, 3(3), 173-179 (2009)
Yasuhisa Kaminaga et al.
The Journal of biological chemistry, 281(33), 23357-23366 (2006-06-13)
We have isolated and characterized Petunia hybrida cv. Mitchell phenylacetaldehyde synthase (PAAS), which catalyzes the formation of phenylacetaldehyde, a constituent of floral scent. PAAS is a cytosolic homotetrameric enzyme that belongs to group II pyridoxal 5'-phosphate-dependent amino-acid decarboxylases and shares
Rajib Saha et al.
PloS one, 6(7), e21784-e21784 (2011-07-15)
The scope and breadth of genome-scale metabolic reconstructions have continued to expand over the last decade. Herein, we introduce a genome-scale model for a plant with direct applications to food and bioenergy production (i.e., maize). Maize annotation is still underway
Fong Lam Chu et al.
Journal of agricultural and food chemistry, 56(22), 10697-10704 (2008-10-29)
Benzaldehyde, a potent aroma chemical of bitter almond, can also be formed thermally from phenylalanine and may contribute to the formation of off-aroma. To identify the precursors involved in its generation during Maillard reaction, various model systems containing phenylalanine, phenylpyruvic
Peter J Landolt et al.
Pest management science, 69(2), 245-249 (2012-08-14)
Phenylacetaldehyde is a flower volatile and attractant for many nectar-seeking moths. Acetic acid is a microbial fermentation product that is present in insect sweet baits. It is weakly attractive to some moths and other insects, but can be additive or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service