Skip to Content
Merck
All Photos(3)

Key Documents

103799

Sigma-Aldrich

Pentafluorophenol

≥99%, for peptide synthesis, ReagentPlus®

Synonym(s):

2,3,4,5,6-Pentafluorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5OH
CAS Number:
Molecular Weight:
184.06
Beilstein:
1912584
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

product name

Pentafluorophenol, ReagentPlus®, ≥99%

product line

ReagentPlus®

Assay

≥99%

form

solid

bp

143 °C (lit.)

mp

34-36 °C (lit.)

application(s)

peptide synthesis

SMILES string

Oc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C6HF5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

XBNGYFFABRKICK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentafluorophenol is widely used as a precursor in both solution and solid-phase peptide synthesis. It is also involved in the preparation of aromatic fluoro derivatives.

Application

Pentafluorophenol can be used as a reactant to synthesize pentafluorophenyl esters from:
  • N-protected amino acid in the presence of dicyclohexylcarbodiimide and ethyl acetate.
  • t-Butoxycarbonyl-L-alanylglycinein the presence of 1-ethyl-(3-dimethylaminopropyl)carbodiimide hydrochloride.
  • Aliphatic dicarboxylic acids in the presence of pyridine, diisopropylcarbodiimide, and acetonitrile.
It can also be used as a reactant to synthesize pentafluorophenyl formate which can be used as a formylating agent in organic synthesis.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentafluorophenol
Jones K
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Yutang Dai et al.
Macromolecular rapid communications, 35(15), 1326-1331 (2014-05-28)
Self-assembly of C3 discotic molecules bearing dipeptide pendants into helical supramolecular polymers is investigated. The dipeptides are constituted from glycine and alanine with altered sequence, aiming at modulating the steric hindrance and examining the steric effects on the assembly. This
Synthesis and Solid-state Conformations of 6S, 8aR/S-6-Alkyl-3, 3-dimethyltetrahydrooxazolo [3, 4-a] pyrazine-5, 8-diones (Pseudoproline Diketopiperazines)
Beauchard A, et al.
Indian Journal of Heterocyclic Chemistry, 28, 11-24 (2018)
Lisbeth Grøndahl et al.
Biomacromolecules, 6(4), 2197-2203 (2005-07-12)
Poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV) melt processed disks and solvent cast films were modified by graft co-polymerization with acrylic acid (AAc) in methanol solution at ambient temperature using gamma irradiation (dose rate of 4.5 kGy/h). To assess the presence of carboxylic acid groups
Benjamin A Ashu-Arrah et al.
Journal of chromatography. A, 1273, 34-43 (2013-01-02)
Pentafluorophenyl and phenyl silica stationary phases offer alternative selectivity compared to alkyl bonded C₁₈ and C₈ stationary phases, through other interactions such as π-π interactions, dipole-dipole and hydrogen bond interactions. Pentafluorophenyl and phenyl silica bonded stationary phases were efficiently prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service