Skip to Content
Merck
All Photos(4)

Key Documents

101303

Sigma-Aldrich

Triphenylmethane

99%

Synonym(s):

1,1′,1′′-Methylidynetris[benzene], Tritane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3CH
CAS Number:
Molecular Weight:
244.33
Beilstein:
1909753
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

358-359 °C/754 mmHg (lit.)

mp

92-94 °C (lit.)

density

1.014 g/mL at 25 °C (lit.)

SMILES string

c1ccc(cc1)C(c2ccccc2)c3ccccc3

InChI

1S/C19H16/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,19H

InChI key

AAAQKTZKLRYKHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chika Takano et al.
Frontiers in microbiology, 8, 1877-1877 (2017-10-20)
Over the past four decades, the incidence of meningitis caused by
Anuradha N Kagalkar et al.
Bioresource technology, 102(22), 10312-10318 (2011-09-20)
Cell cultures of Blumea malcolmii Hook., developed in the laboratory, rapidly decolorized textile industry effluent along with a variety of dyes with diverse structural properties. Most rapid decolorization was observed in case of Malachite Green (93.41% decolorization within 24 h).
Weiguo Huang et al.
Journal of the American Chemical Society, 134(36), 14650-14653 (2012-09-01)
We have increased organic field-effect transistor (OFET) NH(3) response using tris(pentafluorophenyl)borane (TPFB) as a receptor. OFETs with this additive could detect concentrations of 450 ppb v/v, with a limit of detection of 350 ppb, the highest sensitivity reported to date
Marilyn J Schneider et al.
Journal of AOAC International, 98(3), 658-670 (2015-05-31)
A collaborative study was conducted to evaluate the AOAC First Action 2012.25 LC-MS/MS analytical method for the determination of residues of three triphenylmethane dyes (malachite green, crystal violet, and brilliant green) and their metabolites (leucomalachite green and leucocrystal violet) in
Fan Yang et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-07-20)
The development of non-noble metal hydrogen evolution catalysts that can replace Pt is crucial for efficient hydrogen production. Herein, we develop a type of well-dispersed Ni2P on N-doped nanomesh carbon (NC) electrocatalyst by a facile pyrolysis method, which shows excellent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service