HPLC Analysis of Fipronil Enantiomers on Astec® (R,R) P-CAP™
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
Astec (R,R) P-CAP, 25 cm x 4.6 mm I.D., 5 μm particles (31024AST)
mobile phase
[A] heptane; [B] ethanol; (80:20, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 235 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC