Merck
  • Chiral comprehensive two-dimensional gas chromatography with electron-capture detection applied to the analysis of chiral polychlorinated biphenyls in food samples.
Chiral comprehensive two-dimensional gas chromatography with electron-capture detection applied to the analysis of chiral polychlorinated biphenyls in food samples.
Journal of chromatography. A (2005-07-13)
Luisa R Bordajandi, Lourdes Ramos, María José González
ABSTRACT
The feasibility of comprehensive two-dimensional gas chromatography with electron-capture detection (GC x GC-ECD) for the enantioseparation of chiral PCBs from other possible interfering compounds has been evaluated. Three commercially available enantioselective beta-cyclodextrin-based capillary columns (Chirasil-Dex, BGB-172 and BGB-176SE) have been tested as first-dimension columns. Three non-enantioselective stationary phases (HT-8, BPX-50 and Supelcowax-10) were combined with the enantioselective columns to allow the unambiguous determination of the enantiomers of the target chiral PCBs. Each enantioselective first-dimension column tested was able to separate into enantiomers different PCB congeners, but in all cases, the use of Supelcowax-10 as second-dimension column provided the most satisfactory results. The Chirasil-Dex x Supelcowax-10 column combination allowed the determination of the enantiomeric fraction (EF) of PCBs 84, 91, 95, 132, 136, 149, 174 and 176 in the working standard solution, while that of congener 135 was hindered. The BGB-172 x Supelcowax-10 column set allowed a proper EF determination of congeners 45, 84, 131, 132, 135, 171, 174 and 183, while that of PCB 91 was interfered with co-elutants. The column combination BGB-176SE x Supelcowax-10 allowed the determination of all congeners that this enantioselective stationary phase was able to separate into enantiomers, i.e. PCBs 45, 91, 95, 136, 149 and 176. These column combinations have also been evaluated for the simultaneous determination of the 12 congeners with a toxic equivalency factor assigned by the WHO (PCBs 77, 81, 105, 114, 118, 123, 126, 156, 157, 167, 169, 189) and the seven indicator congeners (PCBs 28, 52, 101, 118, 138, 153 and 180), and evaluated for the analysis of food samples.
Product Description
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 2.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 2.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.20 mm, df 0.20 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm