Merck
  • Bidirectional reporter assay using HAL promoter and TOPFLASH improves specificity in high-throughput screening of Wnt inhibitors.
Bidirectional reporter assay using HAL promoter and TOPFLASH improves specificity in high-throughput screening of Wnt inhibitors.
Biotechnology and bioengineering (2017-07-30)
Kiyoshi Yamaguchi, Chi Zhu, Tomoyuki Ohsugi, Yuko Yamaguchi, Tsuneo Ikenoue, Yoichi Furukawa
ABSTRACT
Constitutive activation of Wnt signaling plays an important role in colorectal and liver tumorigenesis. Cell-based assays using synthetic TCF/LEF (T-cell factor/lymphoid enhancer factor) reporters, as readouts of β-catenin/TCF-dependent transcriptional activity, have contributed greatly to the discovery of small molecules that modulate Wnt signaling. In the present study, we report a novel screening method, called a bidirectional dual reporter assay. Integrated transcriptome analysis identified a histidine ammonia-lyase gene (HAL) that was negatively regulated by β-catenin/TCF-dependent transcriptional activity. We leveraged a promoter region of the HAL gene as another transcriptional readout of Wnt signaling. Cells stably expressing both an optimized HAL reporter and the TCF/LEF reporter enabled bidirectional reporter activities in response to Wnt signaling. Increased HAL reporter activity and decreased TCF/LEF reporter activity were observed simultaneously in the cells when β-catenin/TCF7L2 was inhibited. Notably, this method could decrease the number of false positives observed when screening an inhibitor library compared with the conventional TCF/LEF assay. We found that Brefeldin A, a disruptor of the Golgi apparatus, inhibited the Wnt/β-catenin signaling pathway. The utility of our system could be expanded to examine other disease-associated pathways beyond the Wnt/β-catenin signaling pathway.
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Sigma-Aldrich
IWR-1, ≥98% (HPLC)
Sigma-Aldrich
Sulindac sulfide, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-HAL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid