Merck
  • Bioequivalance and pharmacokinetic study of febuxostat in human plasma by using LC-MS/MS with liquid liquid extraction method.
Bioequivalance and pharmacokinetic study of febuxostat in human plasma by using LC-MS/MS with liquid liquid extraction method.
SpringerPlus (2013-06-07)
Babu Rao Chandu, Kanchanamala Kanala, Nagiat T Hwisa, Prakash Katakam, Mukkanti Khagga
ABSTRACT
A bioequivalence study was proved of generic Febuxostat 80 mg tablets (T) in healthy volunteers.For this purpose, Authors developed a simple, sensitive, selective, rapid, rugged and reproducible liquid chromatography-tandem mass spectrometry method for the quantification of Febuxostat (FB) in human plasma using Febuxostat D7 (FBD7) as an internal standard (IS) was used. Chromatographic separation was performed on Ascentis Express C18 (50x4.6 mm, 3.5 μ) column. Mobile phase composed of 10 mM Ammonium formate: Acetonitrile (20:80 v/v), with 0.8 mL/min flow-rate. Drug and IS were extracted by Liquid- liquid extraction. FB and FBD7 were detected with proton adducts at m/z 317.1→261.1 and 324.2→262.1 in multiple reaction monitoring (MRM) positive mode respectively. The method was validated with the correlation coefficients of (r(2)) ≥ 0.9850 over a linear concentration range of 1.00-8000.00 ng/mL. This method demonstrated intra and inter-day precision within 2.64 to 3.88 and 2.76 to 8.44% and accuracy within 97.33 to 99.05 and 100.30 to 103.19% for FB. This method is successfully applied in the Bioequivalence study of 9 human volunteers.
Product Description
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 7.5 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
Ascentis® Express C18, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 2.1 mm